Tisk…

4-Hydroxybenzophenone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (4-Hydroxyphenyl)(phenyl)methanone, p-Hydroxybenzophenone

M-Clarity™ quality level = MQ100

H20202-100G H20202-25G
SIALH20202-100GEA 5970 CZK
SIALH20202-100G SIALH20202-25G
4-Hydroxybenzophenone, Sigma-Aldrich®
4-Hydroxybenzophenone
Vzorec: C₁₃H₁₀O₂
Bod varu: 367 °C (1013 hPa)
Bod tání: 131…133 °C
Hustota: 1,133 g/cm³ (172 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002355
CAS číslo: 1137-42-4
EINECS: 214-507-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR