2-Aminobenzophenone 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
o-Aminobenzophenone
A12580.09
A12580.18
A12580.30
A12580.09EA
1320
CZK
A12580.09
A12580.18
A12580.30
2-Aminobenzophenone 98%
2-Aminobenzophenone
Purity: 98%
CAS No.: 2835-77-0
Molecular Formula: C13H11NO
Formula Weight: 197.24
Flash Point: 240°(464°F)
Melting Point: 105-108°
Boiling Point: 280°
Beilstein Reference: 743425
MDL No.: MFCD00007713
Vzorec:
C₁₃H₁₁NO MW: 197,24 g/mol Bod varu: 280 °C Bod tání: 105…108 °C Hustota: 1,32 g/cm³ (20 °C) Bod vzplanutí: 240 °C (464 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007713 CAS číslo: 2835-77-0 EINECS: 220-613-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...