1,3-Diphenylacetone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1,3-Diphenylpropan-2-one, 1,3-Diphenyl-2-propanone
M-Clarity™ quality level = MQ200
D204609-100G
D204609-25G
SIALD204609-100GEA
4680
CZK
SIALD204609-100G
SIALD204609-25G
1,3-Diphenylacetone, Sigma-Aldrich®
1,3-Diphenylacetone
Vzorec:
(C₆H₅)CH₂C(O)CH₂(C₆H₅) MW: 210,28 g/mol Bod varu: 330 °C (1013 hPa) Bod tání: 30…34 °C Hustota: 1,195 g/cm³ (0 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004795 CAS číslo: 102-04-5 EINECS: 203-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...