Tisk…
4,4-Diphenyl-2-butanone ≥98%
Dodavatel: Thermo Scientific
A13847.03 A13847.06
A13847.03EA 1350 CZK
A13847.03 A13847.06
4,4-Diphenyl-2-butanone ≥98%
4,4-Diphenyl-2-butanone
Purity: 98% CAS No.: 5409-60-9 Molecular Formula: C16H16O Formula Weight: 224.30 MDL No.: MFCD00017619
Vzorec: (C₆H₅)₂CHCH₂C(O)CH₃
MW: 224,3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017619
CAS číslo: 5409-60-9
EINECS: 226-482-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d5ac5e3f946a65cff22c2e8e45aae8c32d1687e&vid=5d5ac5e3f946a65cff22c2e8e45aae8c32d1687e&prid=web&clid=SAPDAM