Tisk…

1,1-Diphenylacetone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: asym. Diphenylacetone, 1,1-Diphenyl-2-propanone

M-Clarity™ quality level = MQ100

D204404-100G D204404-25G
SIALD204404-100GEA 5740 CZK
SIALD204404-100G SIALD204404-25G
1,1-Diphenylacetone, Sigma-Aldrich®
1,1-Diphenylacetone
Vzorec: (C₆H₅)₂CHC(O)CH₃
MW: 210,28 g/mol
Bod varu: 111…126 °C (0,1 mmHg)
Bod tání: 59…62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008759
CAS číslo: 781-35-1
EINECS: 212-307-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7eb18cea6b136cdea03e91f1525c902f74d1578&vid=aabdc13746192c136e72ece83402aebd3b86fb9d&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR