Tisk…

2,2-Diphenylpropiononitrile ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2,2-Diphenylpropionitrile

A17991.06 A17991.09
A17991.06EA 879 CZK
A17991.06 A17991.09
2,2-Diphenylpropiononitrile ≥97%
2,2-Diphenylpropiononitrile

Purity: 97% CAS No.: 5558-67-8 Molecular Formula: C15H13N Formula Weight: 207.28 Flash Point: 110°(230°F) Refractive Index: 1.5720 Boiling Point: 144-145°/2mm Beilstein Reference: 1955952 UN No.: 3276 MDL No.: MFCD00001846

Vzorec: (C₆H₅)₂C(CN)CH₃
MW: 207,27 g/mol
Bod varu: 144…145 °C (2 mmHg)
Hustota: 1,108
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001846
CAS číslo: 5558-67-8
EINECS: 226-925-1
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR