Tisk…

4-Chlorobenzophenone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: p-Chlorobenzophenone

M-Clarity™ quality level = MQ100

C25007-100G C25007-5G
SIALC25007-100GEA 1230 CZK
SIALC25007-100G SIALC25007-5G
4-Chlorobenzophenone, Sigma-Aldrich®
4-Chlorobenzophenone
Vzorec: C₁₃H₉ClO
MW: 216,67 g/mol
Bod varu: 333 °C (1013 hPa)
Bod tání: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00000622
CAS číslo: 134-85-0
EINECS: 205-160-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR