Tisk…

2-Chlorobenzophenone ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: o-Chlorobenzophenone, (2-Chlorophenyl)(phenyl)methanone

A15867.14 A15867.22
A15867.14EA 1500 CZK
A15867.14 A15867.22
2-Chlorobenzophenone ≥99%
2-Chlorobenzophenone

Purity: 99+% CAS No.: 5162-03-8 Molecular Formula: C13H9ClO Formula Weight: 216.67 Flash Point: 110°(230°F) Melting Point: 44-47° Boiling Point: 330° Beilstein Reference: 1869594 MDL No.: MFCD00000558

Vzorec: C₁₃H₉ClO
MW: 216,67 g/mol
Bod varu: 330 °C
Bod tání: 44…47 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000558
CAS číslo: 5162-03-8
EINECS: 225-936-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR