Chloro(diphenyl)methane, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Benzhydryl chloride
M-Clarity™ quality level = MQ200
125032-100G
125032-5G
SIAL125032-100GEA
7050
CZK
SIAL125032-100G
SIAL125032-5G
Chloro(diphenyl)methane, Sigma-Aldrich®
Chloro(diphenyl)methane
Vzorec:
(C₆H₅)₂CHCl MW: 202.68 Bod varu: 315 Bod tání: ∼17 °C Hustota: 1.1346 g/cm³ (20 °C) Bod vzplanutí: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000855 CAS číslo: 90-99-3 EINECS: 202-031-7 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...