Bromkrezolový purpur
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
5',5''-Dibromo-o-cresolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
A19530.09
A19530.18
A19530.09EA
1320
CZK
A19530.09
A19530.18
Bromkrezolový purpur
Bromkrezolový purpur
CAS No.: 115-40-2
Molecular Formula: C21H16Br2O5S
Formula Weight: 540.24
Melting Point: ca 240° dec.
Beilstein Reference: 359618
Merck Reference: 14,1387
MDL No.: MFCD00011681
Vzorec:
C₂₁H₁₆Br₂O₅S Storage Temperature: Ambient |
MDL Number:
MFCD00011681 CAS číslo: 115-40-2 EINECS: 204-087-8 Merck Index: 14,1387 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...