Tetrabromophenol blue
Dodavatel: Thermo Fisher Scientific
Synonyms:
4,5,6,7-Tetrabromo-3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(4,5,6,7-Tetrabromo-3H-2,1-benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide
B20123.03
B20123.06
B20123.14
B20123.03EA
1200
CZK
B20123.03
B20123.06
B20123.14
Tetrabromophenol blue
Tetrabromophenol blue
CAS No.: 4430-25-5
Molecular Formula: C19H6Br8O5S
Formula Weight: 985.55
Melting Point: 203-205° dec.
Beilstein Reference: 378438
MDL No.: MFCD00005876
Vzorec:
C₁₉H₆Br₈O₅S Storage Temperature: Ambient |
MDL Number:
MFCD00005876 CAS číslo: 4430-25-5 EINECS: 224-622-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...