4-Bromobenzophenone ≥98%
Dodavatel: Thermo Fisher Scientific
A14755.14
A14755.22
A14755.30
A14755.14EA
1320
CZK
A14755.14
A14755.22
A14755.30
4-Bromobenzophenone ≥98%
4-Bromobenzophenone
Purity: 98%
CAS No.: 90-90-4
Molecular Formula: C13H9BrO
Formula Weight: 261.12
Odour: Odourless
Melting Point: 79-83°
Boiling Point: 350°
Beilstein Reference: 1910182
MDL No.: MFCD00000103
Vzorec:
C₁₃H₉BrO Bod varu: 350 °C Bod tání: 79…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000103 CAS číslo: 90-90-4 EINECS: 202-024-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...