Tisk…

3-Bromobenzophenone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (3-Bromophenyl)(phenyl)methanone, m-Bromobenzophenone

M-Clarity™ quality level = MQ100

519561-1G 519561-5G
SIAL519561-1GEA 5020 CZK
SIAL519561-1G SIAL519561-5G
3-Bromobenzophenone, Sigma-Aldrich®
3-Bromobenzophenone
Vzorec: C₁₃H₉BrO
MW: 261,12 g/mol
Bod varu: 355 °C (1013 hPa)
Bod tání: 74,5…77.5 °C
Hustota: 1,42 g/cm³ (20 °C)
MDL Number: MFCD00672015
CAS číslo: 1016-77-9
EINECS: 213-808-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR