1-tert-Butyl-4-ethynylbenzene ≥90%
Dodavatel: Thermo Scientific
Varování
|
Synonyms:
1-(tert-Butyl)-4-ethynylbenzene, (4-tert-Butylphenyl)ethyne, (4-tert-Butylphenyl)acetylene, 4-tert-Butylphenylacetylene, 1-Ethynyl-4-tert-butylbenzene
43920.04
43920.09
43920.18
43920.04EA
1370
CZK
43920.04
43920.09
43920.18
1-tert-Butyl-4-ethynylbenzene ≥90%
1-tert-Butyl-4-ethynylbenzene
Purity: 90+%
CAS No.: 772-38-3
Molecular Formula: C12H14
Formula Weight: 158.24
Flash Point: 82°(180°F)
Physical Form: Liquid
Density: 0.878
Refractive Index: 1.5297
Odour: Characteristic
Boiling Point: 130°/40mm
UN No.: 3082
MDL No.: MFCD00190197
Vzorec:
C₁₂H₁₄ MW: 158,24 g/mol Bod varu: 130 °C (40 mmHg) Hustota: 0,878 Bod vzplanutí: 82 °C (180 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190197 CAS číslo: 772-38-3 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...