Octylbenzene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
SIAL113190-5GEA
1310
CZK
SIAL113190-5G
SIAL113190-25G
SIAL113190-100G
Octylbenzene, Sigma-Aldrich®
Octylbenzene
Vzorec:
C₁₄H₂₂ MW: 190,33 g/mol Bod varu: 264 °C (1013 hPa) Bod tání: –3 °C Hustota: 0,8562 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009564 CAS číslo: 2189-60-8 EINECS: 218-582-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...