Tisk…

Sodium toluene-4-sulphonate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: p-Toluenesulphonic acid sodium salt, Sodium p-toluenesulphonate, Toluene-4-sulphonic acid sodium salt

M-Clarity™ quality level = MQ200

152536-100G 152536-500G 152536-5G
SIAL152536-100GEA 2000 CZK
SIAL152536-100G SIAL152536-500G SIAL152536-5G
Sodium toluene-4-sulphonate, Sigma-Aldrich®
Sodium toluene-4-sulphonate
Vzorec: C₇H₇NaO₃S
MW: 194,19 g/mol
MDL Number: MFCD00798566
CAS číslo: 657-84-1
EINECS: 211-522-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR