Ethyl toluene-4-sulphonate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Ethyl 4-toluenesulphonate, Ethyl p-toluenesulphonate, p-Toluenesulphonic acid ethyl ester, Ethyl 4-methylbenzenesulphonate, Toluene-4-sulphonic acid ethyl ester
Purity: 98%
CAS No.: 80-40-0
Molecular Formula: C9H12O3S
Formula Weight: 200.26
Flash Point: 157°(314°F)
Refractive Index: 1.5110
Melting Point: 29-34°
Boiling Point: 221°
Beilstein Reference: 611213
Merck Reference: 14,3858
UN No.: 2811
MDL No.: MFCD00009100
Vzorec:
C₉H₁₂O₃S MW: 200,26 g/mol Bod varu: 221 °C Bod tání: 29…34 °C Hustota: 1,170 Bod vzplanutí: 157 °C (314 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009100 CAS číslo: 80-40-0 EINECS: 201-276-7 UN: 2811 ADR: 6.1,III Merck Index: 14,3858 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...