Toluen-4-sulfonamid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
PTSA, p-Toluenesulphonamide, 4-Methylbenzenesulphonamide, 4-Toluenesulphonamide
M-Clarity™ quality level = MQ100
105902-1KG
105902-250G
SIAL105902-1KGEA
2370
CZK
SIAL105902-1KG
SIAL105902-250G
Toluen-4-sulfonamid, Sigma-Aldrich®
Toluen-4-sulfonamid
Vzorec:
C₇H₉NO₂S Bod varu: 322 °C (1013 hPa) Bod tání: 136…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011692 CAS číslo: 70-55-3 EINECS: 200-741-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...