Toluen-2-sulfonamid ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Methylbenzene-1-sulphonamide, 2-Methylbenzenesulphonamide, o-Toluenesulphonamide
B20544.09
B20544.18
B20544.09EA
1500
CZK
B20544.09
B20544.18
Toluen-2-sulfonamid ≥99%
Toluen-2-sulfonamid
Purity: 99%
CAS No.: 88-19-7
Molecular Formula: C7H9NO2S
Formula Weight: 171.22
Flash Point: 178°(352°F)
Melting Point: 156°
Boiling Point: 210°/1mm
Beilstein Reference: 1102362
MDL No.: MFCD00007934
Vzorec:
C₇H₉NO₂S MW: 171,22 g/mol Bod varu: 210 °C (1 mmHg) Bod tání: 156 °C Bod vzplanutí: 178 °C (352 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007934 CAS číslo: 88-19-7 EINECS: 201-808-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...