m-Toluenesulphonyl chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
3-Toluenesulphonyl chloride, Toluene-3-sulphonyl chloride, Toluene-m-sulphonyl chloride
M-Clarity™ quality level = MQ200
566357-25G
566357-5G
SIAL566357-25GEA
10780
CZK
SIAL566357-25G
SIAL566357-5G
m-Toluenesulphonyl chloride, Sigma-Aldrich®
m-Toluenesulphonyl chloride
Vzorec:
C₇H₇ClO₂S MW: 190,65 g/mol Bod varu: 77…78 °C (0,3 mmHg) Bod tání: 11,7 °C Hustota: 1,314 g/cm³ (20 °C) |
MDL Number:
MFCD00051747 CAS číslo: 1899-93-0 UN: 3265 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...