2-Toluenethiol ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B22460.06
B22460.14
B22460.22
B22460.06EA
1420
CZK
B22460.06
B22460.14
B22460.22
2-Toluenethiol ≥98%
2-Toluenethiol
Purity: 98%
CAS No.: 137-06-4
Molecular Formula: C7H8S
Formula Weight: 124.21
Flash Point: 63°(145°F)
Refractive Index: 1.5800
Melting Point: 10-12°
Boiling Point: 195°
Beilstein Reference: 774068
Merck Reference: 14,9325
UN No.: 2810
MDL No.: MFCD00004838
Vzorec:
C₇H₈S Bod varu: 195 °C Bod tání: 10…12 °C Hustota: 1,054 Bod vzplanutí: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004838 CAS číslo: 137-06-4 EINECS: 205-276-8 UN: 2810 ADR: 6.1,III Merck Index: 14,9325 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...