Tisk…

4-tert-Butylthiophenol ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1-tert-Butyl-4-mercaptobenzene, 4-tert-Butylbenzene-1-thiol, 4-tert-Butylbenzenethiol

A12148.03 A12148.06 A12148.14
A12148.03EA 944 CZK
A12148.03 A12148.06 A12148.14
4-tert-Butylthiophenol ≥97%
4-tert-Butylthiophenol

Purity: 97% CAS No.: 2396-68-1 Molecular Formula: C10H14S Formula Weight: 166.29 Flash Point: 110°(230°F) Refractive Index: 1.5485 Odour: Stench Melting Point: -11° Boiling Point: 238° Beilstein Reference: 606476 MDL No.: MFCD00022067

Vzorec: C₁₀H₁₄S
MW: 166.29
Bod varu: 238 °C
Bod tání: –11 °C
Hustota: 0.990
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00022067
CAS číslo: 2396-68-1
EINECS: 219-255-6
UN: 3334
ADR: 9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR