3-Phenoxytoluene ≥97%
Dodavatel: Thermo Scientific
Synonyms:
Phenyl m-tolyl ether, 1-Methyl-3-phenoxybenzene
B21899.14
B21899.22
B21899.14EA
1800
CZK
B21899.14
B21899.22
3-Phenoxytoluene ≥97%
3-Phenoxytoluene
Purity: 97%
CAS No.: 3586-14-9
Molecular Formula: C13H12O
Formula Weight: 184.24
Flash Point:
Vzorec:
C₁₃H₁₂O MW: 184,24 g/mol Bod varu: 271…273 °C Hustota: 1,052 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008531 CAS číslo: 3586-14-9 EINECS: 222-716-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...