3-Hydroxybenzyl alcohol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(3-Hydroxyphenyl)methanol, KSD 2405, 3-(Hydroxymethyl)phenol
M-Clarity™ quality level = MQ100
H20601-25G
H20601-5G
SIALH20601-25GEA
4600
CZK
SIALH20601-25G
SIALH20601-5G
3-Hydroxybenzyl alcohol, Sigma-Aldrich®
3-Hydroxybenzyl alcohol
Vzorec:
C₇H₈O₂ MW: 124,14 g/mol Bod varu: 86…87 °C (0,001 mmHg) Bod tání: 70 °C Hustota: 1,161 g/cm³ (25 °C) |
MDL Number:
MFCD00004643 CAS číslo: 620-24-6 EINECS: 210-633-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...