2-Methylhydroquinone, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Methylhydroquinone
M-Clarity™ quality level = MQ100
112968-1KG
112968-250G
SIAL112968-1KGEA
6430
CZK
SIAL112968-1KG
SIAL112968-250G
2-Methylhydroquinone, Sigma-Aldrich®
2-Methylhydroquinone
Vzorec:
C₇H₈O₂ MW: 124.14 Bod varu: 285 Bod tání: 123…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002345 CAS číslo: 95-71-6 EINECS: 202-443-7 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...