Tisk…

2-Methylhydroquinone ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Methylhydroquinone

B21241.0B B21241.30
B21241.0BEA 6350 CZK
B21241.0B B21241.30
2-Methylhydroquinone ≥99%
2-Methylhydroquinone

Purity: 99% CAS No.: 95-71-6 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 172°(341°F) Melting Point: 127-130° Boiling Point: 284-286° Beilstein Reference: 2041489 MDL No.: MFCD00002345

Vzorec: C₇H₈O₂
MW: 124.14
Bod varu: 284…286 °C
Bod tání: 127…130 °C
Bod vzplanutí: 172 °C (341 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002345
CAS číslo: 95-71-6
EINECS: 202-443-7
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR