Tisk…

4-Ethylresorcinol ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-Ethylbenzene-1,3-diol

B21575.06 B21575.14 B21575.22
B21575.06EA 1650 CZK
B21575.06 B21575.14 B21575.22
4-Ethylresorcinol ≥98%
4-Ethylresorcinol

Purity: 98% CAS No.: 2896-60-8 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 95-98° Beilstein Reference: 1238231

Vzorec: C₈H₁₀O₂
MW: 138,17 g/mol
Bod varu: 283 °C (1013 hPa)
Bod tání: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00002283
CAS číslo: 2896-60-8
EINECS: 220-777-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR