Tisk…

4-Phenyl-1-butanol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Phenylbutan-1-ol

M-Clarity™ quality level = MQ100

184756-10G 184756-50G
SIAL184756-10GEA 3890 CZK
SIAL184756-10G SIAL184756-50G
4-Phenyl-1-butanol, Sigma-Aldrich®
4-Phenyl-1-butanol
Vzorec: (C₆H₅)(CH₂)₃CH₂OH
MW: 150,22 g/mol
Bod varu: 140 °C (1013 hPa)
Hustota: 0,975 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002971
CAS číslo: 3360-41-6
EINECS: 222-128-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR