3-Phenyl-1-propanol ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-Phenyl-1-Propanol
A13022.0B
A13022.0I
A13022.30
A13022.0BEA
2900
CZK
A13022.0B
A13022.0I
A13022.30
3-Phenyl-1-propanol ≥99%
3-Phenyl-1-propanol
Purity: 99%
CAS No.: 122-97-4
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 109°(228°F)
Refractive Index: 1.5260
Melting Point: -18°
Boiling Point: 235°
Beilstein Reference: 1857542
MDL No.: MFCD00002950
Vzorec:
(C₆H₅)CH₂CH₂CH₂OH Bod varu: 235 °C Bod tání: –18 °C Hustota: 1,004 Bod vzplanutí: 109 °C (228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002950 CAS číslo: 122-97-4 EINECS: 204-587-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...