2-Phenyl-2-propanol ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
α,α-Dimethylbenzyl alcohol
A15580.14
A15580.22
A15580.36
A15580.14EA
2260
CZK
A15580.14
A15580.22
A15580.36
2-Phenyl-2-propanol ≥98%
2-Phenyl-2-propanol
Purity: 98+%
CAS No.: 617-94-7
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 87°(188°F)
Refractive Index: 1.5196
Melting Point: 29-34°
Boiling Point: 215°
Beilstein Reference: 1905012
MDL No.: MFCD00004456
Vzorec:
C₆H₅C(OH)(CH₃)₂ MW: 136,19 g/mol Bod varu: 215 °C Bod tání: 29…34 °C Hustota: 0,973 Bod vzplanutí: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004456 CAS číslo: 617-94-7 EINECS: 210-539-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...