Tisk…

2,4-Dimethoxybenzyl alcohol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

159638-100G 159638-25G
SIAL159638-100GEA 14120 CZK
SIAL159638-100G SIAL159638-25G
2,4-Dimethoxybenzyl alcohol, Sigma-Aldrich®
2,4-Dimethoxybenzyl alcohol
Vzorec: C₉H₁₂O₃
MW: 168,19 g/mol
Bod varu: 317 °C (1013 hPa)
Bod tání: 34…35 °C
MDL Number: MFCD00004614
CAS číslo: 7314-44-5
EINECS: 230-775-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR