Tisk…

3,4-Dimethoxybenzyl alcohol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Veratryl alcohol, (3,4-Dimethoxyphenyl)methanol

M-Clarity™ quality level = MQ100

D133000-25G D133000-100G
SIALD133000-25GEA 1650 CZK
SIALD133000-25G SIALD133000-100G
3,4-Dimethoxybenzyl alcohol, Sigma-Aldrich®
3,4-Dimethoxybenzyl alcohol
Vzorec: C₉H₁₂O₃
MW: 168,19 g/mol
MDL Number: MFCD00004638
CAS číslo: 93-03-8
EINECS: 202-212-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR