4-Hydroxy-3-methoxybenzyl alcohol 99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Vanillyl alcohol, 4-(Hydroxymethyl)-2-methoxyphenol, 2-Hydroxy-5-(hydroxymethyl)anisole, (4-Hydroxy-3-methoxyphenyl)methanol
174580500
174582500
174580010
ACRO174580500EA
1320
CZK
ACRO174580500
ACRO174582500
ACRO174580010
4-Hydroxy-3-methoxybenzyl alcohol 99%
4-Hydroxy-3-methoxybenzyl alcohol
Vzorec:
C₈H₁₀O₃ MW: 154.17 Bod varu: 293 Bod tání: 112…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004659 CAS číslo: 498-00-0 EINECS: 207-852-4 |
Výsledky specifikačního testu
Appearance | Off-white to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 112 to 115 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...