Tisk…

4-Metoxybenzylalkohol, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Anisyl alcohol, p-Anisyl Alcohol, 4-(Hydroxymethyl)anisole, Anise alcohol, (4-Methoxyphenyl)methanol

M-Clarity™ quality level = MQ100

136905-100G 136905-500G 136905-5G
SIAL136905-100GEA 1220 CZK
SIAL136905-100G SIAL136905-500G SIAL136905-5G
4-Metoxybenzylalkohol, Sigma-Aldrich®
4-Metoxybenzylalkohol
Vzorec: C₈H₁₀O₂
MW: 138.17
Bod varu: 145
Bod tání: 20…24 °C
Hustota: 1.1
Storage Temperature: Ambient
MDL Number: MFCD00004653
CAS číslo: 105-13-5
EINECS: 203-273-6
Merck Index: 13,00608

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR