Tisk…

p-Propylanisole, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W293008-10KG-K W293008-1KG-K W293008-5KG-K
SIALW293008-10KG-KEA 46940 CZK
SIALW293008-10KG-K SIALW293008-1KG-K SIALW293008-5KG-K
p-Propylanisole, Sigma-Aldrich®
p-Propylanisole
Vzorec: C₁₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00027121
CAS číslo: 104-45-0
EINECS: 203-203-4
UN: 3082
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR