2-Methylanisole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
o-Cresol methyll ether, 2-Methylanisole, 1-Methoxy-2-methylbenzene, 2-Methoxytoluene
M-Clarity™ quality level = MQ300
W268003-100G-K
W268003-1KG-K
W268003-5KG-K
W268003-SAMPLE
SIALW268003-100G-KEA
0
CZK
SIALW268003-100G-K
SIALW268003-1KG-K
SIALW268003-5KG-K
SIALW268003-SAMPLE
2-Methylanisole, Sigma-Aldrich®
2-Methylanisole
Vzorec:
C₈H₁₀O MW: 122.17 Bod varu: 172 Hustota: 0.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008373 CAS číslo: 578-58-5 EINECS: 209-426-3 UN: 3271 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...