3-Nitro-p-cresol ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
44285.09
44285.18
44285.09EA
2900
CZK
44285.09
44285.18
3-Nitro-p-cresol ≥98%
3-Nitro-p-cresol
Purity: 98%
CAS No.: 2042-14-0
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Physical Form: Powder
Odour: Phenol-like
UN No.: 2446
MDL No.: MFCD00007244
Vzorec:
C₇H₇NO₃ Bod tání: 77…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007244 CAS číslo: 2042-14-0 EINECS: 218-044-6 UN: 2446 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...