4-Methyl-2-nitrophenol ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Nitro-p-cresol, 1-Hydroxy-4-methyl-2-nitrobenzene
A12009.14
A12009.22
A12009.36
A12009.14EA
1460
CZK
A12009.14
A12009.22
A12009.36
4-Methyl-2-nitrophenol ≥97%
4-Methyl-2-nitrophenol
Purity: 97%
CAS No.: 119-33-5
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Flash Point: 108°(226°F)
Odour: Phenol-like
Boiling Point: 125°/22mm
Beilstein Reference: 1868022
MDL No.: MFCD00007120
Vzorec:
C₇H₇NO₃ MW: 153,14 g/mol Bod varu: 125 °C (22 mmHg) Bod tání: 32…35 °C Hustota: 1,240 Bod vzplanutí: 108 °C (226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007120 CAS číslo: 119-33-5 EINECS: 204-315-6 UN: 2446 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...