4-Nitro-m-cresol ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-Methyl-4-nitrophenol, 1-Hydroxy-3-methyl-4-nitrobenzene
L03163.14
L03163.22
L03163.14EA
1250
CZK
L03163.14
L03163.22
4-Nitro-m-cresol ≥98%
4-Nitro-m-cresol
Purity: 98%
CAS No.: 2581-34-2
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Odour: Phenol-like
Melting Point: 126-130°
Beilstein Reference: 1868105
MDL No.: MFCD00007333
Vzorec:
C₇H₇NO₃ MW: 153,14 g/mol Bod tání: 126…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007333 CAS číslo: 2581-34-2 EINECS: 219-952-5 UN: 2446 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...