2,4-Diaminotoluene 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-Methyl-1,3-Phenylenediamine, 1,3-Diamino-4-methylbenzene, Toluene-2,4-diamine
B23495.0B
B23495.30
B23495.0BEA
5220
CZK
B23495.0B
B23495.30
2,4-Diaminotoluene 98%
2,4-Diaminotoluene
Purity: 98%
CAS No.: 95-80-7
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point: 169°(336°F)
Odour: Recognizable
Boiling Point: 283-285°
Beilstein Reference: 2205839
UN No.: 1709
MDL No.: MFCD00007804
Vzorec:
C₇H₁₀N₂ MW: 122,17 g/mol Bod varu: 283…285 °C Bod tání: 96…101 °C Hustota: 1,045 g/cm³ (20 °C) Bod vzplanutí: 169 °C (336 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007804 CAS číslo: 95-80-7 EINECS: 202-453-1 UN: 1709 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...