3,4-Diaminotoluene, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1,2-Diamino-4-methylbenzene, Toluene-3,4-diamine
M-Clarity™ quality level = MQ100
D26024-100G
D26024-5G
SIALD26024-100GEA
1230
CZK
SIALD26024-100G
SIALD26024-5G
3,4-Diaminotoluene, Sigma-Aldrich®
3,4-Diaminotoluene
Vzorec:
C₇H₁₀N₂ MW: 122,17 g/mol Bod varu: 265 °C (1013 hPa) Bod tání: 86…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007728 CAS číslo: 496-72-0 EINECS: 207-826-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...