Tisk…

2,3-Diaminotoluene 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15647.14 A15647.06 A15647.22
A15647.14EA 15510 CZK
A15647.14 A15647.06 A15647.22
2,3-Diaminotoluene 97%
2,3-Diaminotoluene

Purity: 98% CAS No.: 2687-25-4 Molecular Formula: C7H10N2 Formula Weight: 122.17 Melting Point: 56-60° Boiling Point: 225° MDL No.: MFCD00011589

Vzorec: C₇H₁₀N₂
Bod varu: 225 °C
Bod tání: 56…60 °C
Hustota: 1,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011589
CAS číslo: 2687-25-4
EINECS: 220-248-5
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR