Tisk…

4-Aminobenzyl alcohol 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-(Hydroxymethyl)aniline, (4-Aminophenyl)methanol

A14130.06 A14130.14 A14130.22
A14130.06EA 3360 CZK
A14130.06 A14130.14 A14130.22
4-Aminobenzyl alcohol 98%
4-Aminobenzyl alcohol

Purity: 98% CAS No.: 623-04-1 Molecular Formula: C7H9NO Formula Weight: 123.16 Storage Temperature: 2°C to 8°C Melting Point: 59-63° Beilstein Reference: 2078680 MDL No.: MFCD00014782

Vzorec: C₇H₉NO
MW: 123,15 g/mol
Bod varu: 169…171 °C (11 mmHg)
Bod tání: 59…63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014782
CAS číslo: 623-04-1
EINECS: 210-767-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR