Tisk…

2-Aminobenzyl alcohol 98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (2-Aminophenyl)methanol, 2-Hydroxymethylaniline, 2-(Hydroxymethyl)aniline, 2-Aminobenzyl Alcohol

A13180.09 A13180.18 A13180.30
A13180.09EA 834 CZK
A13180.09 A13180.18 A13180.30
2-Aminobenzyl alcohol 98%
2-Aminobenzyl alcohol

Purity: 98% CAS No.: 5344-90-1 Molecular Formula: C7H9NO Formula Weight: 123.16 Storage Temperature: 2°C to 8°C Melting Point: 81-84° Boiling Point: 160°/10mm Beilstein Reference: 1072211 MDL No.: MFCD00007749

Vzorec: C₇H₉NO
Bod varu: 160 °C (10 mmHg)
Bod tání: 81…84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007749
CAS číslo: 5344-90-1
EINECS: 226-293-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR