Tisk…

2-Methoxybenzylamine ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: (2-Methoxyphenyl)methanamine, 2-(Aminomethyl)anisole, o-Anisylamine

L02273.14 L02273.22
L02273.14EA 2590 CZK
L02273.14 L02273.22
2-Methoxybenzylamine ≥98%
2-Methoxybenzylamine

Purity: 98+% CAS No.: 6850-57-3 Molecular Formula: C8H11NO Formula Weight: 137.18 Flash Point: 100°(212°F) Refractive Index: 1.5480 Boiling Point: 228-229° Beilstein Reference: 1100448 UN No.: 2735 MDL No.: MFCD00008110

Vzorec: C₈H₁₁NO
MW: 137,18 g/mol
Bod varu: 228…229 °C
Hustota: 1,057
Bod vzplanutí: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008110
CAS číslo: 6850-57-3
EINECS: 229-941-7
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR