Tisk…

2-Nitro-p-toluidine ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

A13666.0E A13666.22 A13666.36
A13666.0EEA 10020 CZK
A13666.0E A13666.22 A13666.36
2-Nitro-p-toluidine ≥98%
2-Nitro-p-toluidine

Purity: 98+% CAS No.: 89-62-3 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Flash Point: 157°(314°F) Odour: Characteristic Boiling Point: 169°/21mm Beilstein Reference: 879506 UN No.: 2660 MDL No.: MFCD00007907

Vzorec: C₇H₈N₂O₂
Bod varu: 169 °C (21 mmHg)
Bod tání: 115…118 °C
Hustota: 1,31
Bod vzplanutí: 157 °C (314 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007907
CAS číslo: 89-62-3
EINECS: 201-924-9
UN: 2660
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR