Tisk…

5-Nitro-o-toluidine ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

A17726.0E A17726.22 A17726.36
A17726.0EEA 20150 CZK
A17726.0E A17726.22 A17726.36
5-Nitro-o-toluidine ≥98%
5-Nitro-o-toluidine

Purity: 98+% CAS No.: 99-55-8 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Odour: Characteristic Melting Point: 105-108° Beilstein Reference: 879021 UN No.: 2660 MDL No.: MFCD00007741

Vzorec: C₇H₈N₂O₂
Bod tání: 105…108 °C
Hustota: 1,365 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007741
CAS číslo: 99-55-8
EINECS: 202-765-8
UN: 2660
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR