4-Isopropylaniline, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
p-Cumidine, 4-Aminocumene
M-Clarity™ quality level = MQ100
175439-10G
175439-50G
SIAL175439-10GEA
939
CZK
SIAL175439-10G
SIAL175439-50G
4-Isopropylaniline, Sigma-Aldrich®
4-Isopropylaniline
Vzorec:
C₉H₁₃N MW: 135,21 g/mol Bod varu: 226 °C (1013 hPa) Bod tání: –63 °C Hustota: 0,9526 g/cm³ (20 °C) Bod vzplanutí: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007900 CAS číslo: 99-88-7 EINECS: 202-797-2 UN: 1760 ADR: 8,III Merck Index: 13,02647 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...