Tisk…

o-Toluidin ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A13602.0B A13602.0I A13602.30
A13602.0BEA 2010 CZK
A13602.0B A13602.0I A13602.30
o-Toluidin ≥99%
o-Toluidin

Purity: 99% CAS No.: 95-53-4 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 85°(185°F) Refractive Index: 1.5720 Odour: Unpleasant Melting Point: -24° Boiling Point: 199-200° Beilstein Reference: 741981 Merck Reference: 14,9536 UN No.: 1708 MDL No.: MFCD00007730

Vzorec: C₇H₉N
Bod varu: 199…200 °C
Bod tání: –24 °C
Hustota: 1,004
Bod vzplanutí: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007730
CAS číslo: 95-53-4
EINECS: 202-429-0
UN: 1708
ADR: 6.1,II
Merck Index: 14,9536

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR