Tisk…

2-Ethylaniline ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A14984.22 A14984.36
A14984.22EA 862 CZK
A14984.22 A14984.36
2-Ethylaniline ≥97%
2-Ethylaniline

Purity: 97% CAS No.: 578-54-1 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 91°(196°F) Refractive Index: 1.5600 Odour: Characteristic Melting Point: -47° Boiling Point: 208-210° Beilstein Reference: 1841268 UN No.: 2273 MDL No.: MFCD00007751

Vzorec: C₈H₁₁N
Bod varu: 208…210 °C
Bod tání: –47 °C
Hustota: 0,983
Bod vzplanutí: 91 °C (196 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007751
CAS číslo: 578-54-1
EINECS: 209-424-2
UN: 2273
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR